Assay Details
Target Gene Details
Entrez Gene ID: | 729967 |
Gene Name: | MORN repeat containing 2 |
Gene Aliases: |
BLOCK27, MOPT |
Location: |
Chr.2:38875962-38882709 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MORN2 | NM_001145450.1 | 1 | 109 | NP_001138922.1 |
| AY367766.1 | 1 | 109 | AAQ73170.1 | |
| BC102032.1 | AAI02033.1 | |||
| BC102034.1 | AAI02035.1 | |||
| BC102035.1 | AAI02036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2692 | Chr.2:38838626 - 38884730 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map