Assay Details
Target Gene Details
Entrez Gene ID: | 9098 |
Gene Name: | ubiquitin specific peptidase 6 |
Gene Aliases: |
HRP1, TRE17, TRE2, TRESMCR, Tre-2, USP6-short |
Location: |
Chr.17:5116438-5175032 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP6 | NM_001304284.1 | NP_001291213.1 | ||
| NM_004505.3 | NP_004496.2 | |||
| XM_011524050.1 | XP_011522352.1 | |||
| XM_011524051.2 | XP_011522353.1 | |||
| XM_011524052.2 | XP_011522354.1 | |||
| XM_011524053.2 | XP_011522355.1 | |||
| XM_011524054.2 | XP_011522356.1 | |||
| XM_011524055.2 | XP_011522357.1 | |||
| XM_011524056.2 | XP_011522358.1 | |||
| XM_011524057.2 | XP_011522359.1 | |||
| XM_011524058.2 | XP_011522360.1 | |||
| XM_011524059.2 | XP_011522361.1 | |||
| XM_017025290.1 | XP_016880779.1 | |||
| AB449915.1 | ||||
| AY143550.1 | AAN38838.1 | |||
| BX647719.1 | ||||
| X63546.1 | CAA45108.1 | |||
| X63547.1 | CAA45110.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv25210 | Chr.17:5122514 - 5144862 on Build GRCh38 | Gain |
|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv960045 | Chr.17:5124088 - 5179617 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map