Assay Details
Target Gene Details
Entrez Gene ID: | 9913 |
Gene Name: | SPT7-like STAGA complex gamma subunit |
Gene Aliases: |
SPT7L, STAF65, STAF65(gamma), STAF65G, SUPT7H |
Location: |
Chr.2:27642568-27663840 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT7L | NM_001282729.1 | 6 | 2470 | NP_001269658.1 |
| NM_001282730.1 | 6 | 2678 | NP_001269659.1 | |
| NM_001282731.1 | 6 | 2520 | NP_001269660.1 | |
| NM_001282732.1 | 5 | 2060 | NP_001269661.1 | |
| NM_014860.2 | 6 | 2465 | NP_055675.1 | |
| XM_005264672.4 | 6 | 2667 | XP_005264729.1 | |
| AB018307.1 | 6 | 2207 |
More Information
Set Membership: |
|

Genomic Map