Assay Details
Target Gene Details
Entrez Gene ID: | 100137047 |
Gene Name: | jumonji domain containing 7 |
Gene Aliases: |
- |
Location: |
Chr.15:41828085-41837587 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD7 | NM_001114632.1 | 3 | 297 | NP_001108104.1 |
| AK000550.1 | 3 | 403 | ||
| AK000663.1 | 3 | 271 | ||
| AK000814.1 | 2 | 204 | BAA91385.1 | |
| AK225503.1 | 3 | 271 | ||
| AK289833.1 | 3 | 297 | ||
| AK290051.1 | 3 | 266 | ||
| BC025290.1 | 3 | 259 | AAH25290.1 | |
| BC139899.1 | 3 | 276 | ||
| DA100207.1 | 2 | 366 |
Target Gene Details
Entrez Gene ID: | 8681 |
Gene Name: | JMJD7-PLA2G4B readthrough |
Gene Aliases: |
HsT16992, cPLA2-beta |
Location: |
Chr.15:41828085-41848148 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| JMJD7-PLA2G4B | NM_001198588.1 | 3 | 297 | NP_001185517.1 |
| NM_005090.3 | 3 | 297 | NP_005081.1 | |
| AB209150.1 | 3 | 287 | BAD92387.1 | |
| AF065215.1 | 3 | 272 | AAC78836.1 | |
| AF065216.1 | 2 | 348 | ||
| AF121908.1 | 3 | 264 | AAD32135.1 | |
| DA096882.1 | 2 | 373 | ||
| DA121536.1 | 2 | 367 | ||
| DQ523799.1 | 3 | 265 | ||
| DQ523800.1 | 3 | 265 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1035998 | Chr.15:41806698 - 42142056 on Build GRCh38 | Gain |
|
| nsv9244 | Chr.15:41818514 - 41916607 on Build GRCh38 | Gain |
|
| nsv1506 | Chr.15:41824733 - 41857173 on Build GRCh38 | Insertion |
|
| nsv952613 | Chr.15:41816803 - 41884902 on Build GRCh38 | Deletion |
|
| nsv517631 | Chr.15:41804728 - 41895294 on Build GRCh38 | Loss |
|
| dgv2582n100 | Chr.15:41791376 - 41900562 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map