Assay Details
Target Gene Details
Entrez Gene ID: | 64641 |
Gene Name: | early B-cell factor 2 |
Gene Aliases: |
COE2, EBF-2, O/E-3, OE-3 |
Location: |
Chr.8:25841730-26045124 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EBF2 | NM_022659.3 | 16 | 2474 | NP_073150.2 |
| AK021562.1 | 9 | 1513 | BAB13843.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161746 | Chr.8:25843111 - 25850751 on Build GRCh38 | Deletion |
|
| esv2759607 | Chr.8:25823806 - 25849698 on Build GRCh38 | Loss |
|
| esv32961 | Chr.8:25842136 - 25943866 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map