Assay Details
Target Gene Details
Entrez Gene ID: | 93517 |
Gene Name: | short chain dehydrogenase/reductase family 42E, member 1 |
Gene Aliases: |
HSPC105 |
Location: |
Chr.16:81988855-82011488 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SDR42E1 | NM_145168.2 | 3 | 618 | NP_660151.2 |
| XM_005256257.3 | 4 | 823 | XP_005256314.1 | |
| XM_011523471.2 | 3 | 632 | XP_011521773.1 | |
| XM_011523472.2 | 3 | 621 | XP_011521774.1 | |
| AK026740.1 | 3 | 605 | ||
| AK315652.1 | 3 | 579 | ||
| BC019670.1 | 3 | 571 | AAH19670.1 | |
| BX482154.1 |
More Information
Set Membership: |
|

Genomic Map