Assay Details
Target Gene Details
Entrez Gene ID: | 5693 |
Gene Name: | proteasome subunit beta 5 |
Gene Aliases: |
LMPX, MB1, X |
Location: |
Chr.14:23016543-23035220 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PSMB5 | NM_001130725.1 | 2 | 274 | NP_001124197.1 |
| NM_001144932.2 | 2 | 627 | NP_001138404.1 | |
| NM_002797.4 | 2 | 627 | NP_002788.1 | |
| XM_005267871.2 | 2 | 622 | XP_005267928.1 | |
| AK225888.1 | 2 | 392 | ||
| AK300714.1 | 2 | 372 | ||
| AK311895.1 | 2 | 383 | ||
| BC057840.1 | 2 | 382 | AAH57840.1 | |
| BC107720.1 | 2 | 378 | AAI07721.1 | |
| BT006777.1 | 2 | 198 | AAP35423.1 | |
| BU539357.1 | 2 | 250 | ||
| BX248299.1 | 2 | 359 | CAD62626.1 | |
| BX538001.1 | 2 | 374 | CAD97956.1 | |
| CD048996.1 | 2 | 306 | ||
| S74378.1 | 2 | 219 | AAB33092.1 |
More Information
Set Membership: |
|

Genomic Map