Assay Details
Target Gene Details
Entrez Gene ID: | 9682 |
Gene Name: | lysine demethylase 4A |
Gene Aliases: |
JHDM3A, JMJD2, JMJD2A, TDRD14A |
Location: |
Chr.1:43650118-43705518 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4A | NM_014663.2 | 22 | 4330 | NP_055478.2 |
| XM_005271354.3 | 22 | 4367 | XP_005271411.1 | |
| XM_005271355.3 | 22 | 4285 | XP_005271412.1 | |
| XM_005271356.3 | 23 | 4406 | XP_005271413.1 | |
| XM_011542467.2 | 13 | 3438 | XP_011540769.2 | |
| XM_017002912.1 | 23 | 4475 | XP_016858401.1 | |
| AB014577.1 | 22 | 4289 | ||
| CA944984.1 | 1 | 197 |
Target Gene Details
Entrez Gene ID: | 100132774 |
Gene Name: | KDM4A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.1:43699724-43707341 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM4A-AS1 | NR_033827.1 |
More Information
Set Membership: |
|

Genomic Map