Assay Details
Target Gene Details
Entrez Gene ID: | 113451 |
Gene Name: | antizyme inhibitor 2 |
Gene Aliases: |
ADC, AZI2, AZIB1, ODC-p, ODC1L, ODCp |
Location: |
Chr.1:33081110-33162288 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3576805 | Chr.1:32896710 - 33106481 on Build GRCh38 | Gain |
|
| nsv834602 | Chr.1:32947691 - 33116221 on Build GRCh38 | Loss |
|
| nsv545962 | Chr.1:32982619 - 33129611 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs557745273] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map