Assay Details
Target Gene Details
Entrez Gene ID: | 100526693 |
Gene Name: | ARPC4-TTLL3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.3:9792548-9836356 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARPC4-TTLL3 | NM_001198793.1 | NP_001185722.1 | ||
| AK024110.1 | BAB14828.1 |
Target Gene Details
Entrez Gene ID: | 26140 |
Gene Name: | tubulin tyrosine ligase like 3 |
Gene Aliases: |
HOTTL |
Location: |
Chr.3:9809960-9836356 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTLL3 | NM_001025930.3 | NP_001021100.3 | ||
| NR_037162.1 | ||||
| AF078842.1 | AAF23353.1 | |||
| AK023960.1 | BAB14741.1 | |||
| AK125875.1 | ||||
| AK297289.1 | ||||
| AK315140.1 | ||||
| AL833939.1 | CAD38794.1 | |||
| BP357921.1 | ||||
| BX440835.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
| esv3595249 | Chr.3:9812974 - 9821465 on Build GRCh38 | Gain |
|
| nsv819644 | Chr.3:9817550 - 9818201 on Build GRCh38 | Gain |
|
| nsv834615 | Chr.3:9791053 - 9971639 on Build GRCh38 | Loss |
|
| nsv237238 | Chr.3:9816535 - 9822062 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map