Assay Details
Target Gene Details
Entrez Gene ID: | 8669 |
Gene Name: | eukaryotic translation initiation factor 3 subunit J |
Gene Aliases: |
EIF3S1, eIF3-alpha, eIF3-p35 |
Location: |
Chr.15:44537068-44562803 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J | NM_001284335.1 | 7 | 1976 | NP_001271264.1 |
| NM_001284336.1 | 6 | 1991 | NP_001271265.1 | |
| NM_003758.3 | 8 | 2138 | NP_003749.2 | |
| AK002006.1 | 8 | 2058 | ||
| BU621047.1 | 1 | 437 | ||
| U97670.1 | 9 | 2005 | AAC78729.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs141191303] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map