Assay Details
Target Gene Details
Entrez Gene ID: | 55198 |
Gene Name: | adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2 |
Gene Aliases: |
DIP13B |
Location: |
Chr.12:105173297-105236230 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| APPL2 | NM_001251904.1 | 21 | 2696 | NP_001238833.1 |
| NM_001251905.1 | 21 | 2721 | NP_001238834.1 | |
| NM_018171.3 | 21 | 2678 | NP_060641.2 | |
| XM_006719472.1 | 20 | 2474 | XP_006719535.1 | |
| XM_011538530.2 | 21 | 4148 | XP_011536832.1 | |
| XM_011538531.2 | 21 | 3811 | XP_011536833.1 | |
| XM_011538532.2 | 21 | 5137 | XP_011536834.1 | |
| XM_017019551.1 | 21 | 3083 | XP_016875040.1 | |
| XM_017019552.1 | 21 | 3793 | XP_016875041.1 | |
| XM_017019553.1 | 22 | 5391 | XP_016875042.1 | |
| XM_017019554.1 | 20 | 2456 | XP_016875043.1 | |
| AK001521.2 | 21 | 2498 | ||
| AY113704.1 | 21 | 2678 | AAM55530.1 | |
| BC033731.1 | 21 | 2582 | AAH33731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832506 | Chr.12:105067496 - 105243783 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77220426] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map