Assay Details
Target Gene Details
Entrez Gene ID: | 1739 |
Gene Name: | discs large MAGUK scaffold protein 1 |
Gene Aliases: |
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg |
Location: |
Chr.3:197042560-197299272 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLG1 | NM_001098424.1 | 1 | 61 | NP_001091894.1 |
| NM_001204386.1 | 1 | 61 | NP_001191315.1 | |
| NM_001290983.1 | NP_001277912.1 | |||
| NM_004087.2 | 1 | 61 | NP_004078.2 | |
| XM_017005801.1 | XP_016861290.1 | |||
| XM_017005803.1 | XP_016861292.1 | |||
| XM_017005808.1 | XP_016861297.1 | |||
| XM_017005809.1 | 1 | 100 | XP_016861298.1 | |
| XM_017005810.1 | XP_016861299.1 | |||
| XM_017005813.1 | XP_016861302.1 | |||
| XM_017005814.1 | 1 | 101 | XP_016861303.1 | |
| XM_017005817.1 | XP_016861306.1 | |||
| XM_017005818.1 | 1 | 72 | XP_016861307.1 | |
| XM_017005819.1 | XP_016861308.1 | |||
| AB855790.1 | ||||
| AB855791.1 | ||||
| AK304280.1 | 1 | 61 | ||
| AK310596.1 | ||||
| BC140841.1 | ||||
| BC144651.1 | ||||
| EF553524.1 | ||||
| U13896.1 | 1 | 61 | AAA50598.1 | |
| U13897.1 | 1 | 61 | AAA50599.1 |
Target Gene Details
Entrez Gene ID: | 100507086 |
Gene Name: | DLG1 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.3:197298247-197303751 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DLG1-AS1 | NR_038289.1 | 1 | 270 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| nsv829830 | Chr.3:197161008 - 197354023 on Build GRCh38 | Gain |
|
| esv3893852 | Chr.3:197204950 - 197333165 on Build GRCh38 | Gain |
|
| nsv428431 | Chr.3:197128182 - 197316896 on Build GRCh38 | Gain+Loss |
|
| esv3599304 | Chr.3:197291633 - 197300133 on Build GRCh38 | Gain |
|
| nsv516873 | Chr.3:197066987 - 197300056 on Build GRCh38 | Gain+Loss |
|
| dgv5038n100 | Chr.3:197134594 - 197659420 on Build GRCh38 | Gain |
|
| nsv593116 | Chr.3:197183211 - 197617305 on Build GRCh38 | Gain |
|
| nsv818205 | Chr.3:197291254 - 197300056 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map