Assay Details
Target Gene Details
Entrez Gene ID: | 1893 |
Gene Name: | extracellular matrix protein 1 |
Gene Aliases: |
URBWD |
Location: |
Chr.1:150508011-150513789 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECM1 | NM_001202858.1 | NP_001189787.1 | ||
| NM_004425.3 | NP_004416.2 | |||
| NM_022664.2 | NP_073155.2 | |||
| AK097046.1 | ||||
| AK097205.1 | 3 | 1827 | ||
| AK292435.1 | ||||
| AK301369.1 | 6 | 809 | ||
| AK302279.1 | ||||
| AK316390.1 | 6 | 788 | ||
| BC023505.2 | AAH23505.1 | |||
| U65932.1 | AAB05933.1 | |||
| U68186.1 | AAB88081.1 | |||
| U68187.1 | AAB88082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1006838 | Chr.1:150453465 - 150750307 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map