Assay Details
Target Gene Details
Entrez Gene ID: | 92304 |
Gene Name: | secretoglobin family 3A member 1 |
Gene Aliases: |
HIN-1, HIN1, LU105, PnSP-2, UGRP2 |
Location: |
Chr.5:180590105-180591487 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCGB3A1 | NM_052863.2 | NP_443095.2 | ||
| AF086152.1 | ||||
| AF436839.1 | AAQ04481.1 | |||
| AY040564.1 | AAK82942.1 | |||
| AY359064.1 | AAQ89423.1 | |||
| BC029176.1 | AAH29176.1 | |||
| BC072673.1 | AAH72673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2762572 | Chr.5:180569987 - 180756359 on Build GRCh38 | Gain |
|
| nsv1026417 | Chr.5:180294376 - 180864360 on Build GRCh38 | Gain |
|
| nsv509111 | Chr.5:180577379 - 180654864 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map