Assay Details
Target Gene Details
Entrez Gene ID: | 6464 |
Gene Name: | SHC adaptor protein 1 |
Gene Aliases: |
SHC, SHCA |
Location: |
Chr.1:154962298-154974492 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHC1 | NM_001130040.1 | 12 | 2484 | NP_001123512.1 |
| NM_001130041.1 | 13 | 2169 | NP_001123513.1 | |
| NM_001202859.1 | 13 | 2169 | NP_001189788.1 | |
| NM_003029.4 | 13 | 2172 | NP_003020.2 | |
| NM_183001.4 | 12 | 2481 | NP_892113.4 | |
| XM_011509892.2 | 12 | 3724 | XP_011508194.1 | |
| XM_011509893.2 | 12 | 3722 | XP_011508195.1 | |
| XM_011509894.2 | 11 | 3674 | XP_011508196.1 | |
| XM_011509897.1 | 12 | 2109 | XP_011508199.1 | |
| XM_017002081.1 | 11 | 3642 | XP_016857570.1 | |
| XM_017002082.1 | 11 | 3640 | XP_016857571.1 | |
| XM_017002083.1 | 12 | 2079 | XP_016857572.1 | |
| AB208849.1 | 8 | 3209 | BAD92086.1 | |
| AK096169.1 | 12 | 2467 | ||
| AK296786.1 | ||||
| AK315842.1 | ||||
| BC014158.2 | 13 | 2053 | AAH14158.1 | |
| BC033925.1 | 10 | 1883 | AAH33925.1 | |
| BX647136.1 | 13 | 2397 | ||
| BX647149.1 | 12 | 2484 | ||
| U73377.1 | 12 | 2454 | AAB49972.1 | |
| X68148.1 | 13 | 2011 | CAA48251.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv509502 | Chr.1:154904407 - 155142291 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map