Assay Details
Target Gene Details
Entrez Gene ID: | 10044 |
Gene Name: | SH2 domain containing 3C |
Gene Aliases: |
CHAT, NSP3, PRO34088, SHEP1 |
Location: |
Chr.9:127738317-127778769 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH2D3C | NM_001142531.1 | 11 | 2027 | NP_001136003.1 |
| NM_001142532.1 | 11 | 2276 | NP_001136004.1 | |
| NM_001142533.1 | 10 | 2164 | NP_001136005.1 | |
| NM_001142534.1 | 10 | 2136 | NP_001136006.1 | |
| NM_001252334.1 | 12 | 2533 | NP_001239263.1 | |
| NM_005489.3 | 11 | 2162 | NP_005480.2 | |
| NM_170600.2 | 12 | 2635 | NP_733745.1 | |
| XM_005251639.1 | 11 | 2143 | XP_005251696.1 | |
| XM_011518114.1 | 12 | 2619 | XP_011516416.1 | |
| XM_011518115.1 | 12 | 2579 | XP_011516417.1 | |
| XM_011518117.2 | 10 | 3255 | XP_011516419.1 | |
| XM_017014174.1 | 10 | 2416 | XP_016869663.1 | |
| AF124251.1 | 11 | 2074 | AAD28246.1 | |
| AI472242.1 | ||||
| AJ420431.1 | 5 | 704 | ||
| AK056068.1 | 10 | 2164 | ||
| AK096983.1 | 8 | 1457 | ||
| AK128287.1 | 13 | 2566 | ||
| AK291393.1 | 10 | 2128 | ||
| AK292041.1 | 11 | 2090 | ||
| AK292718.1 | 12 | 2506 | ||
| AK294386.1 | 11 | 2027 | ||
| AY358089.1 | 11 | 2144 | AAQ88456.1 | |
| AY358440.1 | 11 | 2063 | AAQ89948.1 | |
| BC027962.1 | 11 | 2033 | AAH27962.1 | |
| BC032365.1 | 12 | 2510 | AAH32365.1 | |
| BX647905.1 | 12 | 2459 | CAI46101.1 | |
| CU454732.1 | 4 | 511 | ||
| CU455802.1 | 4 | 511 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12879n54 | Chr.9:127731234 - 127824409 on Build GRCh38 | Loss |
|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|
| nsv471323 | Chr.9:127727464 - 127845239 on Build GRCh38 | Loss |
|
| nsv466573 | Chr.9:127735841 - 127787855 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map