Assay Details
Target Gene Details
Entrez Gene ID: | 4084 |
Gene Name: | MAX dimerization protein 1 |
Gene Aliases: |
BHLHC58, MAD, MAD1 |
Location: |
Chr.2:69915041-69942945 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXD1 | NM_001202513.1 | 1 | 296 | NP_001189442.1 |
| NM_001202514.1 | 1 | 296 | NP_001189443.1 | |
| NM_002357.3 | 1 | 296 | NP_002348.1 | |
| AK301088.1 | 1 | 228 | ||
| AK312734.1 | 1 | 228 | ||
| BC069377.1 | 1 | 90 | AAH69377.1 | |
| BC069433.1 | 1 | 90 | AAH69433.1 | |
| BC098396.1 | 1 | 266 | AAH98396.1 | |
| BC113531.1 | 1 | 149 | ||
| BC117260.1 | 1 | 149 | ||
| BC143831.1 | 1 | 149 | ||
| BC143832.1 | 1 | 149 | ||
| CR536495.1 | CAG38734.1 | |||
| CR541692.1 | CAG46493.1 | |||
| DB445256.1 | 1 | 291 | ||
| DC347823.1 | 1 | 296 | ||
| L06895.1 | 1 | 153 | AAA36194.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834254 | Chr.2:69745440 - 69921396 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map