Assay Details
Target Gene Details
Entrez Gene ID: | 2045 |
Gene Name: | EPH receptor A7 |
Gene Aliases: |
EHK-3, EHK3, EK11, HEK11 |
Location: |
Chr.6:93240020-93419687 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHA7 | NM_001288629.1 | 17 | 4145 | NP_001275558.1 |
| NM_004440.3 | 17 | 4160 | NP_004431.1 | |
| XM_005248669.2 | 17 | 4205 | XP_005248726.1 | |
| XM_017010365.1 | 17 | 4190 | XP_016865854.1 | |
| L36642.1 | 17 | 4104 | AAA74243.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604163 | Chr.6:93232143 - 93380358 on Build GRCh38 | Gain |
|
| nsv517329 | Chr.6:92538625 - 94814709 on Build GRCh38 | Gain+Loss |
|
| nsv604162 | Chr.6:93227679 - 93268190 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map