Assay Details
Target Gene Details
Entrez Gene ID: | 6372 |
Gene Name: | C-X-C motif chemokine ligand 6 |
Gene Aliases: |
CKA-3, GCP-2, GCP2, SCYB6 |
Location: |
Chr.4:73836556-73838760 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL6 | NM_002993.3 | NP_002984.1 | ||
| U81234.1 | AAD00506.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
| nsv4389 | Chr.4:73824830 - 73856555 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map