Assay Details
Target Gene Details
Entrez Gene ID: | 3490 |
Gene Name: | insulin like growth factor binding protein 7 |
Gene Aliases: |
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF |
Location: |
Chr.4:57031071-57110385 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGFBP7 | NM_001553.2 | 5 | 910 | NP_001544.1 |
| AI761388.1 | 1 | 228 | ||
| BC017201.2 | 5 | 898 | AAH17201.1 | |
| BC066339.1 | 5 | 910 | AAH66339.1 | |
| BT006654.1 | AAP35300.1 | |||
| L19182.1 | 5 | 890 | AAA16187.1 | |
| S75725.1 | 5 | 898 | AAB32370.1 |
More Information
Set Membership: |
|

Genomic Map