Assay Details
Target Gene Details
Entrez Gene ID: | 94134 |
Gene Name: | Rho GTPase activating protein 12 |
Gene Aliases: |
- |
Location: |
Chr.10:31805398-31928876 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP12 | NM_001270695.1 | 19 | 3282 | NP_001257624.1 |
| NM_001270696.1 | 18 | 3207 | NP_001257625.1 | |
| NM_001270697.1 | 19 | 3156 | NP_001257626.1 | |
| NM_001270698.1 | 17 | 3066 | NP_001257627.1 | |
| NM_001270699.1 | 16 | 2908 | NP_001257628.1 | |
| NM_018287.6 | 20 | 3297 | NP_060757.4 | |
| XM_005252644.1 | 18 | 3141 | XP_005252701.1 | |
| XM_011519761.1 | 18 | 3075 | XP_011518063.1 | |
| XM_017016954.1 | 16 | 2985 | XP_016872443.1 | |
| AF131804.1 | 2 | 726 | ||
| AK000744.1 | 16 | 2350 | ||
| AK001833.1 | 3 | 1065 | BAA91932.1 | |
| AK025438.1 | 13 | 1708 | BAB15131.1 | |
| AK091182.1 | 18 | 3152 | ||
| AL137485.1 | 11 | 1735 | CAB70766.1 | |
| AL834250.2 | 16 | 2832 | CAD38926.2 | |
| AY033594.1 | 18 | 3174 | AAK52311.1 | |
| AY033595.1 | 20 | 3264 | AAK52312.1 | |
| BC051811.1 | 8 | 1319 | AAH51811.1 | |
| BC094719.1 | 19 | 3206 | AAH94719.1 | |
| BX641115.1 | 2 | 2651 | CAE46053.1 | |
| CR936681.1 |
More Information
Set Membership: |
|

Genomic Map