Assay Details
Target Gene Details
Entrez Gene ID: | 550 |
Gene Name: | ancient ubiquitous protein 1 |
Gene Aliases: |
- |
Location: |
Chr.2:74526648-74529897 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AUP1 | NM_181575.4 | 2 | 377 | NP_853553.1 |
| NR_126510.1 | 2 | 377 | ||
| NR_126511.1 | 2 | 377 | ||
| XM_005264392.3 | 2 | 243 | XP_005264449.1 | |
| AF100746.1 | 2 | 150 | AAD43010.1 | |
| AF100753.1 | 2 | 177 | AAD43017.1 | |
| AF100754.1 | 2 | 177 | AAD43018.1 | |
| AF165515.1 | 2 | 157 | AAF86645.1 | |
| AK023983.1 | 2 | 155 | BAB14753.1 | |
| AK055566.1 | 2 | 197 | ||
| AK293121.1 | 2 | 196 | ||
| AK301398.1 | 2 | 176 | ||
| AK304886.1 | 2 | 240 | ||
| AK308231.1 | 2 | 178 | ||
| AK310159.1 | 2 | 178 | ||
| BC033646.2 | 2 | 192 | ||
| CR749661.1 | 2 | 169 | CAH18454.2 | |
| CR999537.1 | 2 | 363 | ||
| CX164419.1 | 2 | 159 | ||
| HY078082.1 | 2 | 378 |
Target Gene Details
Entrez Gene ID: | 27429 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
OMI, PARK13, PRSS25 |
Location: |
Chr.2:74529405-74533556 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HTRA2 | NM_001321727.1 | NP_001308656.1 | ||
| NM_001321728.1 | NP_001308657.1 | |||
| NM_013247.4 | NP_037379.1 | |||
| NM_145074.2 | NP_659540.1 | |||
| NR_135769.1 | ||||
| NR_135770.1 | ||||
| NR_135771.1 | ||||
| NR_135772.1 | ||||
| AF141305.1 | AAF66596.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458185 | Chr.2:74367887 - 74531807 on Build GRCh38 | Loss |
|
| esv2759061 | Chr.2:74252797 - 74535298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map