Assay Details
Target Gene Details
Entrez Gene ID: | 286464 |
Gene Name: | cilia and flagella associated protein 47 |
Gene Aliases: |
CHDC2, CXorf22, CXorf30, CXorf59 |
Location: |
Chr.X:35919734-36385319 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP47 | NM_001304548.1 | 8 | 1340 | NP_001291477.1 |
| NM_152632.3 | 8 | 1340 | NP_689845.2 | |
| XM_017029452.1 | 8 | 1333 | XP_016884941.1 | |
| XM_017029453.1 | 8 | 1333 | XP_016884942.1 | |
| AK126295.1 | 8 | 1306 | BAC86517.1 |
More Information
Set Membership: |
|

Genomic Map