Assay Details
Target Gene Details
Entrez Gene ID: | 56287 |
Gene Name: | gastrokine 1 |
Gene Aliases: |
AMP18, BRICD1, CA11, FOV, foveolin |
Location: |
Chr.2:68974573-68980980 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GKN1 | NM_019617.3 | 6 | 624 | NP_062563.3 |
| AB039886.1 | 6 | 624 | BAA92433.1 | |
| AJ966789.1 | 6 | 624 | CAI85015.1 | |
| AL833013.1 | 1 | 2226 | CAH56295.1 | |
| AY139184.1 | 6 | 562 | AAN75449.1 | |
| AY359050.1 | 6 | 548 | AAQ89409.1 | |
| AY456961.1 | 6 | 561 | AAR21211.1 | |
| BC059778.1 | 6 | 550 |
More Information
Set Membership: |
|

Genomic Map