Assay Details
Target Gene Details
Entrez Gene ID: | 84166 |
Gene Name: | NLR family CARD domain containing 5 |
Gene Aliases: |
CLR16.1, NOD27, NOD4 |
Location: |
Chr.16:56989547-57083524 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRC5 | NM_032206.4 | 2 | 268 | NP_115582.4 |
| XM_005256193.2 | 3 | 329 | XP_005256250.1 | |
| XM_005256194.3 | 3 | 540 | XP_005256251.1 | |
| XM_005256195.2 | 3 | 329 | XP_005256252.1 | |
| XM_005256196.2 | 3 | 329 | XP_005256253.1 | |
| XM_005256197.2 | 3 | 329 | XP_005256254.1 | |
| XM_005256199.3 | 3 | 329 | XP_005256256.1 | |
| XM_005256201.3 | 3 | 329 | XP_005256258.1 | |
| XM_006721297.2 | 3 | 317 | XP_006721360.1 | |
| XM_006721298.2 | 3 | 329 | XP_006721361.1 | |
| XM_006721300.2 | 3 | 329 | XP_006721363.1 | |
| XM_011523373.2 | 3 | 476 | XP_011521675.1 | |
| XM_011523375.2 | 3 | 329 | XP_011521677.1 | |
| XM_017023766.1 | 3 | 329 | XP_016879255.1 | |
| XM_017023767.1 | 3 | 317 | XP_016879256.1 | |
| XM_017023768.1 | 3 | 329 | XP_016879257.1 | |
| XM_017023769.1 | 3 | 317 | XP_016879258.1 | |
| XM_017023770.1 | 3 | 329 | XP_016879259.1 | |
| XM_017023771.1 | 3 | 329 | XP_016879260.1 | |
| AF389420.1 | 1 | 141 | AAO59377.1 | |
| AK074182.1 | 2 | 281 | BAB85008.1 | |
| AK097030.1 | 3 | 318 | ||
| AL704292.1 | 3 | 366 | ||
| DB293035.1 | 3 | 401 | ||
| EF452236.1 | 1 | 141 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1059098 | Chr.16:56733938 - 57028336 on Build GRCh38 | Gain |
|
| nsv827683 | Chr.16:57016822 - 57147283 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map