Assay Details
Target Gene Details
Entrez Gene ID: | 54902 |
Gene Name: | tetratricopeptide repeat domain 19 |
Gene Aliases: |
2010204O13Rik, MC3DN2 |
Location: |
Chr.17:15999380-16045428 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC19 | NM_001271420.1 | 1 | 267 | NP_001258349.1 |
| NM_017775.3 | 1 | 267 | NP_060245.3 | |
| XM_017024801.1 | 1 | 180 | XP_016880290.1 | |
| XM_017024802.1 | 1 | 180 | XP_016880291.1 | |
| AK056878.1 | 1 | 179 | ||
| BC105128.1 | 1 | 267 | AAI05129.1 | |
| BC112107.1 | 1 | 267 | AAI12108.1 |
Target Gene Details
Entrez Gene ID: | 125150 |
Gene Name: | zinc finger SWIM-type containing 7 |
Gene Aliases: |
SWS1 |
Location: |
Chr.17:15976560-15999692 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZSWIM7 | NM_001042697.1 | 1 | 47 | NP_001036162.1 |
| NM_001042698.1 | 1 | 47 | NP_001036163.1 | |
| BU075283.1 | 1 | 50 | ||
| DQ530600.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv3108n100 | Chr.17:15966243 - 16193927 on Build GRCh38 | Gain |
|
| esv3892973 | Chr.17:15947882 - 16030711 on Build GRCh38 | Gain |
|
| esv3572431 | Chr.17:15951820 - 16194089 on Build GRCh38 | Gain |
|
| esv29086 | Chr.17:15999311 - 16001396 on Build GRCh38 | Loss |
|
| dgv3107n100 | Chr.17:15703953 - 16150198 on Build GRCh38 | Gain |
|
| esv2715702 | Chr.17:15750514 - 16668268 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map