Assay Details
Target Gene Details
Entrez Gene ID: | 81501 |
Gene Name: | dendrocyte expressed seven transmembrane protein |
Gene Aliases: |
FIND, TM7SF4, hDC-STAMP |
Location: |
Chr.8:104339087-104356874 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCSTAMP | NM_001257317.1 | NP_001244246.1 | ||
| NM_030788.3 | 3 | 1197 | NP_110415.1 | |
| XM_011517321.1 | 4 | 1566 | XP_011515623.1 | |
| XM_011517324.1 | XP_011515626.1 | |||
| XM_017013878.1 | 5 | 1665 | XP_016869367.1 | |
| XM_017013879.1 | XP_016869368.1 | |||
| XM_017013880.1 | XP_016869369.1 | |||
| AF277290.1 | 3 | 1175 | AAL02152.1 | |
| AF305068.1 | 3 | 1169 | AAG39167.1 | |
| AK095417.1 | 3 | 1167 | ||
| BC069349.1 | 3 | 1167 | AAH69349.1 | |
| BC112018.1 | 2 | 1141 | AAI12019.1 | |
| BC112020.1 | 2 | 1141 | AAI12021.1 | |
| BC171732.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1032775 | Chr.8:104299136 - 104397744 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map