Assay Details
Target Gene Details
Entrez Gene ID: | 51006 |
Gene Name: | solute carrier family 35 member C2 |
Gene Aliases: |
BA394O2.1, C20orf5, CGI-15, OVCOV1 |
Location: |
Chr.20:46349528-46364461 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057686 | Chr.20:46105657 - 46434770 on Build GRCh38 | Loss |
|
| nsv586155 | Chr.20:46340501 - 46387008 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map