Assay Details
Target Gene Details
Entrez Gene ID: | 23528 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
ZBP-99, ZNP-99 |
Location: |
Chr.1:200404947-200483604 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF281 | NM_001281293.1 | 2 | 3398 | NP_001268222.1 |
| NM_001281294.1 | 3 | 3290 | NP_001268223.1 | |
| NM_012482.4 | 2 | 3394 | NP_036614.1 | |
| XM_017000887.1 | 5 | 4779 | XP_016856376.1 | |
| XM_017000888.1 | 5 | 3737 | XP_016856377.1 | |
| AK021707.1 | 1 | 1697 | ||
| AK022921.1 | 1 | 2024 | ||
| BC060820.1 | 2 | 3374 | AAH60820.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv548990 | Chr.1:200400630 - 200452338 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map