Assay Details
Target Gene Details
Entrez Gene ID: | 10675 |
Gene Name: | chondroitin sulfate proteoglycan 5 |
Gene Aliases: |
NGC |
Location: |
Chr.3:47562238-47580240 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSPG5 | NM_001206942.1 | 5 | 2118 | NP_001193871.1 |
| NM_001206943.1 | 5 | 2121 | NP_001193872.1 | |
| NM_001206944.1 | 4 | 1964 | NP_001193873.1 | |
| NM_001206945.1 | 5 | 2199 | NP_001193874.1 | |
| NM_006574.3 | 5 | 2040 | NP_006565.2 | |
| AF461087.1 | 5 | 3963 | AAQ04774.1 | |
| AF461088.1 | 4 | 1972 | AAQ04775.1 | |
| AF461089.1 | 5 | 4044 | AAQ04776.1 | |
| AI989800.1 | 1 | 124 | ||
| AK094639.1 | 1 | 2403 | ||
| BM674119.1 | 1 | 136 | ||
| BU732090.1 | 1 | 136 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3807 | Chr.3:47558395 - 47607814 on Build GRCh38 | Insertion |
|
| nsv428417 | Chr.3:46638962 - 47778557 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map