Assay Details
Target Gene Details
Entrez Gene ID: | 79660 |
Gene Name: | protein phosphatase 1 regulatory subunit 3B |
Gene Aliases: |
GL, PPP1R4, PTG |
Location: |
Chr.8:9136254-9151642 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R3B | NM_001201329.1 | 2 | 1641 | NP_001188258.1 |
| NM_024607.3 | 2 | 1493 | NP_078883.2 | |
| XM_006716253.3 | 2 | 1446 | XP_006716316.1 | |
| XM_017013835.1 | 2 | 1593 | XP_016869324.1 | |
| AK024067.1 | 2 | 1479 | BAB14811.1 | |
| AK096119.1 | 2 | 1573 | ||
| BC043388.1 | 2 | 1426 | AAH43388.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv610206 | Chr.8:9103507 - 9304538 on Build GRCh38 | Loss |
|
| nsv436801 | Chr.8:9140246 - 9140696 on Build GRCh38 | Insertion |
|
| esv3616157 | Chr.8:8901663 - 9165103 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map