Assay Details
Target Gene Details
Entrez Gene ID: | 22949 |
Gene Name: | prostaglandin reductase 1 |
Gene Aliases: |
LTB4DH, PGR1, ZADH3 |
Location: |
Chr.9:111549722-111599855 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGR1 | NM_001146108.1 | NP_001139580.1 | ||
| NM_001146109.1 | NP_001139581.1 | |||
| NM_012212.3 | NP_036344.2 | |||
| XM_011518394.2 | XP_011516696.1 | |||
| XM_011518395.2 | XP_011516697.1 | |||
| XM_017014485.1 | XP_016869974.1 | |||
| AA876375.1 | ||||
| AK091948.1 | 11 | 2128 | ||
| AK289597.1 | ||||
| AK298379.1 | ||||
| BC035228.2 | AAH35228.1 | |||
| D49387.1 | BAA08382.1 |
Target Gene Details
Entrez Gene ID: | 158399 |
Gene Name: | zinc finger protein 483 |
Gene Aliases: |
ZKSCAN16, ZSCAN48 |
Location: |
Chr.9:111525159-111577844 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF483 | NM_001007169.3 | NP_001007170.1 | ||
| AK092811.1 | BAC03981.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621491 | Chr.9:111562503 - 111563727 on Build GRCh38 | Loss |
|
| esv2678607 | Chr.9:111562478 - 111563764 on Build GRCh38 | Deletion |
|
| nsv1054737 | Chr.9:111562245 - 111612449 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map