Assay Details
Target Gene Details
Entrez Gene ID: | 160492 |
Gene Name: | lamin tail domain containing 1 |
Gene Aliases: |
IFLTD1, PAS1C1 |
Location: |
Chr.12:25476082-25648604 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMNTD1 | NM_001145728.2 | 10 | 1643 | NP_001139200.1 |
| NM_001256266.1 | 10 | 1555 | NP_001243195.1 | |
| NM_152590.3 | 9 | 1524 | NP_689803.2 | |
| XM_011520575.1 | 12 | 1749 | XP_011518877.1 | |
| AK093323.1 | 9 | 1485 | BAC04132.1 | |
| AK302292.1 | ||||
| BC037957.1 | 10 | 1524 | AAH37957.1 | |
| DW009862.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76519300] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map