Assay Details
Target Gene Details
Entrez Gene ID: | 8208 |
Gene Name: | chromatin assembly factor 1 subunit B |
Gene Aliases: |
CAF-1, CAF-IP60, CAF1, CAF1A, CAF1P60, MPHOSPH7, MPP7 |
Location: |
Chr.21:36380145-36416827 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHAF1B | NM_005441.2 | NP_005432.1 | ||
| XM_011529755.2 | XP_011528057.1 | |||
| XM_017028477.1 | XP_016883966.1 | |||
| XM_017028478.1 | XP_016883967.1 | |||
| AK313152.1 | ||||
| AL711314.1 | ||||
| BC021218.2 | AAH21218.1 | |||
| U20980.1 | AAA76737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33578 | Chr.21:36116014 - 36447329 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map