Assay Details
Target Gene Details
Entrez Gene ID: | 132949 |
Gene Name: | aminoadipate-semialdehyde dehydrogenase |
Gene Aliases: |
ACSF4, LYS2, NRPS1098, NRPS998 |
Location: |
Chr.4:56337687-56387677 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AASDH | NM_001286668.1 | 14 | 3015 | NP_001273597.1 |
| NM_001286669.1 | 14 | 3166 | NP_001273598.1 | |
| NM_001286670.1 | 12 | 2690 | NP_001273599.1 | |
| NM_001286671.1 | 12 | 2868 | NP_001273600.1 | |
| NM_001323890.1 | 16 | 3404 | NP_001310819.1 | |
| NM_001323892.1 | 14 | 3123 | NP_001310821.1 | |
| NM_001323893.1 | 13 | 3002 | NP_001310822.1 | |
| NM_001323899.1 | 14 | 3180 | NP_001310828.1 | |
| NM_181806.3 | 15 | 3287 | NP_861522.2 | |
| XM_017007740.1 | 14 | 3116 | XP_016863229.1 | |
| XM_017007741.1 | 16 | 3363 | XP_016863230.1 | |
| XM_017007742.1 | 13 | 3063 | XP_016863231.1 | |
| XM_017007743.1 | 13 | 2952 | XP_016863232.1 | |
| XM_017007744.1 | 12 | 2870 | XP_016863233.1 | |
| AA743758.1 | ||||
| AF516672.1 | 14 | 2930 | AAP76519.1 | |
| AK294332.1 | 12 | 2652 | ||
| AK303733.1 | ||||
| AK304309.1 | 15 | 3278 | ||
| AK316296.1 | 15 | 3268 | ||
| AY314787.1 | 15 | 3250 | AAQ83120.1 | |
| AY422212.1 | 15 | 3202 | AAR31184.1 | |
| BC071815.1 | 1 | 201 | AAH71815.1 | |
| BC142709.1 | 12 | 2804 | ||
| BX640635.1 | 10 | 4118 | CAH56482.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161086 | Chr.4:56310907 - 56510457 on Build GRCh38 | Duplication |
|
| nsv829940 | Chr.4:56206175 - 56396564 on Build GRCh38 | Gain |
|
| dgv5213n100 | Chr.4:56076751 - 56538345 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map