Assay Details
Target Gene Details
Entrez Gene ID: | 23435 |
Gene Name: | TAR DNA binding protein |
Gene Aliases: |
ALS10, TDP-43 |
Location: |
Chr.1:11012622-11030528 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARDBP | NM_007375.3 | 6 | 3744 | NP_031401.1 |
| XM_017000863.1 | 6 | 3732 | XP_016856352.1 | |
| XM_017000864.1 | XP_016856353.1 | |||
| XM_017000865.1 | XP_016856354.1 | |||
| XM_017000866.1 | 7 | 2998 | XP_016856355.1 | |
| XM_017000867.1 | XP_016856356.1 | |||
| XM_017000868.1 | XP_016856357.1 | |||
| AF311304.1 | 1 | 642 | AAG47782.1 | |
| BM716882.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832425 | Chr.1:10843788 - 11062918 on Build GRCh38 | Loss |
|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114897688] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map