Assay Details
Target Gene Details
Entrez Gene ID: | 23635 |
Gene Name: | single stranded DNA binding protein 2 |
Gene Aliases: |
HSPC116, SOSS-B2 |
Location: |
Chr.5:81417360-81751253 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3605594 | Chr.5:81406976 - 81473957 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map