Assay Details
Target Gene Details
Entrez Gene ID: | 29760 |
Gene Name: | B-cell linker |
Gene Aliases: |
AGM4, BASH, BLNK-S, LY57, SLP-65, SLP65, bca |
Location: |
Chr.10:96189171-96271576 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
| nsv831947 | Chr.10:96085409 - 96258168 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map