Assay Details
Target Gene Details
Entrez Gene ID: | 122011 |
Gene Name: | casein kinase 1 alpha 1 like |
Gene Aliases: |
CK1 |
Location: |
Chr.13:37103260-37105664 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSNK1A1L | NM_145203.5 | 1 | 641 | NP_660204.2 |
| BC028723.2 | 1 | 643 | AAH28723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv826612 | Chr.13:37104536 - 37106155 on Build GRCh38 | Loss |
|
| esv3892330 | Chr.13:36864962 - 37227904 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map