Assay Details
Target Gene Details
Entrez Gene ID: | 80014 |
Gene Name: | WW and C2 domain containing 2 |
Gene Aliases: |
BOMB |
Location: |
Chr.4:183098677-183320777 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2 | XM_011532269.2 | XP_011530571.1 |
Target Gene Details
Entrez Gene ID: | 152641 |
Gene Name: | WWC2 antisense RNA 2 |
Gene Aliases: |
C4orf38 |
Location: |
Chr.4:183097021-183099199 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WWC2-AS2 | NR_024008.1 | 1 | 527 | |
| AK054839.1 | 1 | 527 | BAB70812.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv596340 | Chr.4:183098699 - 183100083 on Build GRCh38 | Gain+Loss |
|
| esv26194 | Chr.4:183098192 - 183100106 on Build GRCh38 | Loss |
|
| nsv822872 | Chr.4:183097250 - 183100919 on Build GRCh38 | Gain |
|
| nsv1016376 | Chr.4:182859067 - 183262599 on Build GRCh38 | Gain |
|
| dgv9415n54 | Chr.4:183095835 - 183099302 on Build GRCh38 | Loss |
|
| dgv9416n54 | Chr.4:183098458 - 183099974 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map