Assay Details
Target Gene Details
Entrez Gene ID: | 23085 |
Gene Name: | ELKS/RAB6-interacting/CAST family member 1 |
Gene Aliases: |
Cast2, ELKS, ERC-1, RAB6IP2 |
Location: |
Chr.12:990010-1495933 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2251n54 | Chr.12:553282 - 1101718 on Build GRCh38 | Gain |
|
| nsv556943 | Chr.12:997617 - 1111522 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map