Assay Details
Target Gene Details
Entrez Gene ID: | 6660 |
Gene Name: | SRY-box 5 |
Gene Aliases: |
L-SOX5, L-SOX5B, L-SOX5F, LAMSHF |
Location: |
Chr.12:23529499-24562701 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
More Information
Set Membership: |
|

Genomic Map