Assay Details
Target Gene Details
Entrez Gene ID: | 23259 |
Gene Name: | DDHD domain containing 2 |
Gene Aliases: |
SAMWD1, SPG54, iPLA(1)gamma |
Location: |
Chr.8:38231343-38273442 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDHD2 | NM_001164232.1 | NP_001157704.1 | ||
| NM_015214.2 | NP_056029.2 | |||
| XM_005273454.2 | XP_005273511.1 | |||
| XM_005273455.3 | XP_005273512.1 | |||
| XM_005273456.3 | XP_005273513.1 | |||
| XM_011544456.1 | XP_011542758.1 | |||
| XM_017013252.1 | XP_016868741.1 | |||
| XM_017013253.1 | XP_016868742.1 | |||
| XM_017013254.1 | XP_016868743.1 | |||
| XM_017013255.1 | XP_016868744.1 | |||
| XM_017013256.1 | XP_016868745.1 | |||
| AB018268.1 | BAA34445.1 | |||
| AK023218.1 | BAB14470.1 | |||
| AK056525.1 | ||||
| AK125904.1 | ||||
| AK127040.1 | ||||
| AK299875.1 | ||||
| BC010504.1 | AAH10504.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7162n100 | Chr.8:38159377 - 38249049 on Build GRCh38 | Gain |
|
| dgv7165n100 | Chr.8:38200929 - 38249049 on Build GRCh38 | Loss |
|
| nsv1029521 | Chr.8:38236953 - 38249049 on Build GRCh38 | Gain |
|
| nsv1020415 | Chr.8:38223888 - 38249049 on Build GRCh38 | Gain+Loss |
|
| dgv7164n100 | Chr.8:38194164 - 38310837 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map