Assay Details
Target Gene Details
Entrez Gene ID: | 79165 |
Gene Name: | leukocyte receptor cluster member 1 |
Gene Aliases: |
- |
Location: |
Chr.19:54155639-54159743 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LENG1 | NM_024316.2 | 4 | 681 | NP_077292.2 |
| AF211966.1 | 3 | 459 | AAG01393.1 | |
| BC014986.1 | 4 | 646 | AAH14986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644783 | Chr.19:54151022 - 54159664 on Build GRCh38 | Gain |
|
| nsv515623 | Chr.19:54102901 - 54179150 on Build GRCh38 | Loss |
|
| nsv953613 | Chr.19:54097214 - 54198733 on Build GRCh38 | Deletion |
|
| esv2718812 | Chr.19:53184636 - 54322450 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map