Assay Details
Target Gene Details
Entrez Gene ID: | 79109 |
Gene Name: | mitogen-activated protein kinase associated protein 1 |
Gene Aliases: |
JC310, MIP1, SIN1, SIN1b, SIN1g |
Location: |
Chr.9:125437394-125707234 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAPKAP1 | NM_001006617.1 | 12 | 2403 | NP_001006618.1 |
| NM_001006619.1 | 11 | 2262 | NP_001006620.1 | |
| NM_001006620.1 | 11 | 2075 | NP_001006621.1 | |
| NM_001006621.1 | 10 | 1985 | NP_001006622.1 | |
| NM_024117.3 | 11 | 2295 | NP_077022.1 | |
| XM_011519005.1 | 13 | 2544 | XP_011517307.1 | |
| XM_011519007.1 | 12 | 2403 | XP_011517309.1 | |
| XM_017015126.1 | 12 | 2292 | XP_016870615.1 | |
| AK295364.1 | 9 | 1709 | ||
| AL833042.1 | 11 | 2036 | CAH56311.1 | |
| AY524429.1 | 12 | 2400 | AAS90839.1 | |
| AY524431.1 | 11 | 2292 | AAS90841.1 | |
| AY524432.1 | 11 | 2259 | AAS90842.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831722 | Chr.9:125302099 - 125476079 on Build GRCh38 | Gain |
|
| nsv1046626 | Chr.9:125359313 - 125470470 on Build GRCh38 | Gain |
|
| nsv515979 | Chr.9:125388505 - 125465747 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map