Assay Details
Target Gene Details
Entrez Gene ID: | 100507577 |
Gene Name: | uncharacterized LOC100507577 |
Gene Aliases: |
- |
Location: |
Chr.16:48355685-48362999 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507577 | NR_040677.1 | |||
| AK056051.1 |
Target Gene Details
Entrez Gene ID: | 6477 |
Gene Name: | siah E3 ubiquitin protein ligase 1 |
Gene Aliases: |
SIAH1A |
Location: |
Chr.16:48360531-48448440 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIAH1 | NM_001006610.1 | 2 | 2246 | NP_001006611.1 |
| NM_003031.3 | 2 | 1874 | NP_003022.3 | |
| XM_006721246.1 | 3 | 2050 | XP_006721309.1 | |
| XM_011523279.1 | 6 | 3390 | XP_011521581.1 | |
| BC042550.1 | 2 | 2216 | AAH42550.1 | |
| BC044920.1 | AAH44920.1 | |||
| BX647064.1 | 1 | 2818 | CAE46191.1 | |
| U63295.1 | 1 | 1757 | AAC12950.1 | |
| U76247.1 | 2 | 1943 | AAC51907.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758645 | Chr.16:48295526 - 48445843 on Build GRCh38 | Gain |
|
| nsv833219 | Chr.16:48256684 - 48425390 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map