Assay Details
Target Gene Details
Entrez Gene ID: | 7485 |
Gene Name: | tryptophan rich basic protein |
Gene Aliases: |
CHD5, GET1 |
Location: |
Chr.21:39380287-39397889 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WRB | NM_004627.4 | 1 | 92 | NP_004618.2 |
| AK293113.1 | ||||
| AK293866.1 | ||||
| AK297186.1 | ||||
| AK299245.1 | 1 | 53 | ||
| BC012415.1 | AAH12415.1 | |||
| DB500750.1 | 1 | 92 | ||
| Y12478.1 | CAA73081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4433n100 | Chr.21:39369967 - 39418271 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map