Assay Details
Target Gene Details
Entrez Gene ID: | 55818 |
Gene Name: | lysine demethylase 3A |
Gene Aliases: |
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA |
Location: |
Chr.2:86440647-86492716 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM3A | NM_001146688.1 | 5 | 558 | NP_001140160.1 |
| NM_018433.5 | 5 | 827 | NP_060903.2 | |
| XM_006712051.3 | 4 | 530 | XP_006712114.1 | |
| XM_017004493.1 | 5 | 846 | XP_016859982.1 | |
| XM_017004494.1 | 5 | 616 | XP_016859983.1 | |
| AB018285.2 | 5 | 590 | ||
| AK304276.1 | 3 | 344 | ||
| AL832150.1 | 5 | 593 | CAH18459.3 | |
| BG723267.1 | 5 | 610 | ||
| BX640698.1 | 5 | 628 | CAE45820.1 | |
| BX647271.1 | 5 | 596 | ||
| BX647752.1 | 5 | 656 | ||
| BX648841.1 | 5 | 559 | ||
| BX649109.1 | 5 | 597 | ||
| CR749581.1 | 5 | 595 | CAH18373.1 |
More Information
Set Membership: |
|

Genomic Map