Assay Details
Target Gene Details
Entrez Gene ID: | 285429 |
Gene Name: | DDB1 and CUL4 associated factor 4 like 1 |
Gene Aliases: |
WDR21B |
Location: |
Chr.4:41981696-41986467 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF4L1 | NM_001029955.3 | 1 | 1997 | NP_001025126.2 |
| AK122914.1 | 1 | 1915 | ||
| BC035027.1 | 1 | 1870 | AAH35027.1 | |
| BC052943.1 | 1 | 1937 | AAH52943.1 |

Genomic Map